Learn More
TARGETMOL CHEMICALS INC Mitoxantrone 200MG
SDP
Catalog No. 502880047
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Mitoxantrone (mitozantrone) is an anthracenedione antibiotic with antineoplastic activity. Mitoxantrone intercalates into and crosslinks DNA, thereby disrupting DNA and RNA replication. This agent also binds to topoisomerase II, resulting in DNA strand breaks and inhibition of DNA repair. Mitoxantrone is less cardiotoxic compared to doxorubicin. Purity 98.74%
Catalog No. 50-288-0047 Supplier TARGETMOL CHEMICALS INC Supplier No. T6588200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Mitoxantrone (mitozantrone) is an anthracenedione antibiotic with antineoplastic activity. Mitoxantrone intercalates into and crosslinks DNA, thereby disrupting DNA and RNA replication. This agent also binds to topoisomerase II, resulting in DNA strand breaks and inhibition of DNA repair. Mitoxantrone is less cardiotoxic compared to doxorubicin. Purity 98.74%