Learn More
Selleck Chemical LLC Mitoxantrone 2HCl S2485-1g
SDP
Catalog No. 503111632
Encompass_Preferred
Mitoxantrone 2HCl is a dihydrochloride salt of Mitoxantrone Mitoxantrone is an inhibitor of type II topoisomerase and protein kinase C (PKC) with IC50 of 8 5 M for PKC Mitoxantrone inhibits cell proliferative growth of MCF-7/wt cells with IC50 of 0 42 M Mitoxantrone also induces apoptosis
Catalog No. 50-311-1632 Supplier Selleck Chemical LLC Supplier No. S24851G
Mitoxantrone 2HCl is a dihydrochloride salt of Mitoxantrone Mitoxantrone is an inhibitor of type II topoisomerase and protein kinase C (PKC) with IC50 of 8 5 M for PKC Mitoxantrone inhibits cell proliferative growth of MCF-7/wt cells with IC50 of 0 42 M Mitoxantrone also induces apoptosis