Learn More
Selleck Chemical LLC Mitoxantrone 2HCl 50mg 70476-82-3 NSC-301739
SDP
Catalog No. 501362459
Encompass_Preferred
Mitoxantrone 2HCl (NSC-301739) is a dihydrochloride salt of Mitoxantrone. Mitoxantrone is an inhibitor of type II topoisomerase and protein kinase C (PKC) with IC50 of 8.5 ?M for PKC. Mitoxantrone inhibits cell proliferative growth of MCF-7/wt cells with IC50 of 0.42 ?M. Mitoxantrone also induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2459 Supplier Selleck Chemical LLC Supplier No. S248550MG
Mitoxantrone 2HCl (NSC-301739) is a dihydrochloride salt of Mitoxantrone. Mitoxantrone is an inhibitor of type II topoisomerase and protein kinase C (PKC) with IC50 of 8.5 ?M for PKC. Mitoxantrone inhibits cell proliferative growth of MCF-7/wt cells with IC50 of 0.42 ?M. Mitoxantrone also induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.