Learn More
Medchemexpress LLC HY-13502A 100mg , Mitoxantrone (dihydrochloride) mitozantrone dihydrochloride CAS:70476-82-3 Purity:98%
SDP
Catalog No. 501493381
Encompass_Preferred
Medchemexpress, HY-13502A 100mg Mitoxantrone (dihydrochloride) mitozantrone dihydrochloride CAS:70476-82-3 Formula:C22H30Cl2N4O6 IC50: 8.5 μM (PKC) Purity:98% Mitoxantrone dihydrochloride is a topoisomerase II inhibitor; also inhibits protein kinase C ( PKC ) activity with an IC 50 of 8.5 μM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3381 Supplier Medchemexpress LLC Supplier No. HY13502A100MG
Medchemexpress, HY-13502A 100mg Mitoxantrone (dihydrochloride) mitozantrone dihydrochloride CAS:70476-82-3 Formula:C22H30Cl2N4O6 IC50: 8.5 μM (PKC) Purity:98% Mitoxantrone dihydrochloride is a topoisomerase II inhibitor; also inhibits protein kinase C ( PKC ) activity with an IC 50 of 8.5 μM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.