Learn More
Abcam Mito x antrone dihydrochloride, Type II topoisomerase inhibitor, 50MG
Catalog No. 502706954
Encompass_Preferred
MW 517.4 Da, Purity >98%. Type II topoisomerase inhibitor (IC₅₀ = 50 nM). Able to inhibit DNA and protein synthesis, and induce both DNA strand breakage and cross linkage. Antineoplastic, immunomodulatory and chemotherapeutic action.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6954 Supplier Abcam Supplier No. AB14104150MG
MW 517.4 Da, Purity >98%. Type II topoisomerase inhibitor (IC₅₀ = 50 nM). Able to inhibit DNA and protein synthesis, and induce both DNA strand breakage and cross linkage. Antineoplastic, immunomodulatory and chemotherapeutic action.
The product is subject to the following: Abcam Restricted Use Statement