Learn More
Cayman Chemical Mitoxantrone (hydrochloride), Purity: ≥95%, CAS: 70476-82-3, 50 mg
SDP
Catalog No. 502968804
Encompass_Preferred
An inhibitor of DNA topoisomerase IIα and HIV-1 integrase (IC50s = 5.3 and 3.8 μM, respectively); intercalates DNA in a cell-free assay at 2 µM; inhibits DNA synthesis and induces DNA-protein crosslinks in vitro; inhibits the growth of K562 and K562-derived etoposide-resistant K/VP.5 cells (IC50s = 0.42 and 1.68 µM, respectively); inhibits HIV-1 integrase (IC50 = 3.8 μM); inhibits the membrane fusion of a mixed population of HEK293T cells expressing either ACE2 or the extracellular domain SARS-CoV-2 spike glycoprotein; inhibits the entry of SARS-CoV-2 into Vero E6 cells at 0.2 µM
Catalog No. 50-296-8804 Supplier Cayman Chemical Supplier No. 1484250 MG
An inhibitor of DNA topoisomerase IIα and HIV-1 integrase (IC50s = 5.3 and 3.8 μM, respectively); intercalates DNA in a cell-free assay at 2 µM; inhibits DNA synthesis and induces DNA-protein crosslinks in vitro; inhibits the growth of K562 and K562-derived etoposide-resistant K/VP.5 cells (IC50s = 0.42 and 1.68 µM, respectively); inhibits HIV-1 integrase (IC50 = 3.8 μM); inhibits the membrane fusion of a mixed population of HEK293T cells expressing either ACE2 or the extracellular domain SARS-CoV-2 spike glycoprotein; inhibits the entry of SARS-CoV-2 into Vero E6 cells at 0.2 µM