Learn More
Bioworld Mitra Maintenance/ Replate Medium, 25 L
Catalog No. 50254992
Encompass_Preferred
Mitra Maintenance/ Replate MediumContains the Macronutrients, Micronutrients, Vitamins, and Supplements as described by Mitra et al. (1976) with addition of Activated Charcoal, Agar, & SucroseCytokines and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 29.82 g/LPartially soluble in water
Catalog No. 50-254-992 Supplier Bioworld Supplier No. 306301973
Mitra Maintenance/ Replate MediumContains the Macronutrients, Micronutrients, Vitamins, and Supplements as described by Mitra et al. (1976) with addition of Activated Charcoal, Agar, & SucroseCytokines and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 29.82 g/LPartially soluble in water