Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mitraphylline | 509-80-8 | 10 MG
SDP

Catalog No. 5000418354
Encompass_Preferred

Mitraphylline is the major pentacyclic oxindolic alkaloid found in Uncaria tomentosa. It inhibits lipopolysaccharide-mediated activation of primary human neutrophils and exhibits antiproliferative activity in human glioma and neuroblastoma cell lines. Mitraphylline can also inhibit interleukin release in vivo.

  • Purity: 99.30%
  • Appearance: solid, white to off-white
  • Solubility (in vitro): 6.76 mg/mL in DMSO (18.35 mM); requires ultrasonic
  • Storage: 4°C, sealed, away from moisture and light
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture and light)
  • Reduces NO production and downregulates pro-inflammatory cytokines in human LPS-activated neutrophils
  • Exhibits antiproliferative activity in GAMG (IC50 = 20.05 μM) and SKN-BE cells
  • Impairs TNF-α liberation and inhibits IL-1α/β in mice in vivo
  • For research use only

Catalog No. 50-004-18354 Supplier Medchemexpress LLC Supplier No. HYN694610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mitraphylline is the major pentacyclic oxindolic alkaloid found in Uncaria tomentosa. It inhibits lipopolysaccharide-mediated activation of primary human neutrophils and exhibits antiproliferative activity in human glioma and neuroblastoma cell lines. Mitraphylline can also inhibit interleukin release in vivo.

  • Purity: 99.30%
  • Appearance: solid, white to off-white
  • Solubility (in vitro): 6.76 mg/mL in DMSO (18.35 mM); requires ultrasonic
  • Storage: 4°C, sealed, away from moisture and light
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture and light)
  • Reduces NO production and downregulates pro-inflammatory cytokines in human LPS-activated neutrophils
  • Exhibits antiproliferative activity in GAMG (IC50 = 20.05 μM) and SKN-BE cells
  • Impairs TNF-α liberation and inhibits IL-1α/β in mice in vivo
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.