Learn More
Cayman Chemical MItrOS580 500test
SDP
Catalog No. 502934682
Encompass_Preferred
A fluorescent probe for mitochondrial superoxide production in live cells; cell-permeable and selectively targets mitochondria; upon oxidation by superoxide, MitoROS™ 580 fluoresces, and this fluorescence can be measured using fluorescence microscopy or flow cytometry; ex/em = 510/580 nm; has been used to determine effects of curcumin treatment on superoxide production in Jurkat cells
Catalog No. 50-293-4682 Supplier Cayman Chemical Supplier No. 25169500 TEST
A fluorescent probe for mitochondrial superoxide production in live cells; cell-permeable and selectively targets mitochondria; upon oxidation by superoxide, MitoROS™ 580 fluoresces, and this fluorescence can be measured using fluorescence microscopy or flow cytometry; ex/em = 510/580 nm; has been used to determine effects of curcumin treatment on superoxide production in Jurkat cells