Learn More
Med Vet International Open Palm X-ray Mitt, J0677P
Catalog No. 5000495089
Encompass_Preferred
Shields the back of the hand and features hook and loop strap closure across the palm for quick removal. Covered in dark blue weblon. Standard protection: 0.5mm Pb equivalency.
Catalog No. 50-004-95089 Supplier Med Vet International Supplier No. JOR677P
Shields the back of the hand and features hook and loop strap closure across the palm for quick removal. Covered in dark blue weblon. Standard protection: 0.5mm Pb equivalency.