Learn More
MOLTOX Molecular Toxicology Inc Mitus Salivarius Agar, 20 mL/plate, 10 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948813
Encompass_Preferred
Mitis Salivarius Agar is used in conjunction with 1% Tellurite solution for isolating Streptococcus mitis, S. salivarius and enterococci from contaminated specimens. These organisms are commonly found in the flora of the oral cavity. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-813 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 5140S013
Mitis Salivarius Agar is used in conjunction with 1% Tellurite solution for isolating Streptococcus mitis, S. salivarius and enterococci from contaminated specimens. These organisms are commonly found in the flora of the oral cavity. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.