Learn More
MSC Mitutoyo ABSOLUTE Digimatic Depth Gage, 0-8 in, 0.001 in Accuracy, Stainless Steel
Catalog No. 5000534602
Encompass
This depth gage provides accurate and reliable measurements for various industrial and laboratory applications. Constructed from durable stainless steel, it offers a measurement range of 0 to 8 inches (0 to 200 mm) and includes advanced functionalities for efficient operation.
  • Origin set function
  • Zero setting for precise measurements
  • Automatic power on/off conserves battery life
  • Inch/mm conversion for flexible unit display
  • SPC data output for statistical process control
  • Equipped with a long-life SR44 battery
  • Base and measuring faces are hardened and micro-lapped for durability
Catalog No. 50-005-34602 Supplier MSC Supplier No. 75267906
This depth gage provides accurate and reliable measurements for various industrial and laboratory applications. Constructed from durable stainless steel, it offers a measurement range of 0 to 8 inches (0 to 200 mm) and includes advanced functionalities for efficient operation.
  • Origin set function
  • Zero setting for precise measurements
  • Automatic power on/off conserves battery life
  • Inch/mm conversion for flexible unit display
  • SPC data output for statistical process control
  • Equipped with a long-life SR44 battery
  • Base and measuring faces are hardened and micro-lapped for durability