Learn More
MSC Mitutoyo ABSOLUTE Digimatic Electronic Caliper, 4 in, 100 mm, 0.01 mm Resolution
Catalog No. 5000516919
Encompass
The ABSOLUTE Digimatic Caliper features an electromagnetic induction type absolute encoder and AOS (Advanced Onsite Sensor). This design ensures precise measurements by eliminating origin setting after power-on and providing smooth, stable operation for various applications.
  • Absolute scale eliminates the need for origin setting after power-on.
  • Smooth, stable, and comfortable operation with an ergonomic slider.
  • Long battery life of approximately 3.5 years.
  • Step-measurement capable.
  • High-contrast LCD with 9mm font height reduces user fatigue.
  • AOS sensor repels dirt, oil, and water, ensuring accuracy.
  • ABS and INC measurement functions.
  • Includes inside diameter jaws, depth measurement rod, battery, and case.
  • Constructed from hardened stainless steel.
Catalog No. 50-005-16919 Supplier MSC Supplier No. 06257950
The ABSOLUTE Digimatic Caliper features an electromagnetic induction type absolute encoder and AOS (Advanced Onsite Sensor). This design ensures precise measurements by eliminating origin setting after power-on and providing smooth, stable operation for various applications.
  • Absolute scale eliminates the need for origin setting after power-on.
  • Smooth, stable, and comfortable operation with an ergonomic slider.
  • Long battery life of approximately 3.5 years.
  • Step-measurement capable.
  • High-contrast LCD with 9mm font height reduces user fatigue.
  • AOS sensor repels dirt, oil, and water, ensuring accuracy.
  • ABS and INC measurement functions.
  • Includes inside diameter jaws, depth measurement rod, battery, and case.
  • Constructed from hardened stainless steel.