Learn More
MSC Mitutoyo Blade Micrometer, Mechanical, 1 to 2 in, 0.000150 in Accuracy
Catalog No. 5000533357
Encompass
This mechanical blade micrometer offers precise measurements within a 1.0000 to 2.0000 inch range and an accuracy of 0.000150 " It features a ratchet thimble for consistent pressure and an IP54 rating for water resistance, making it suitable for various demanding environments.
  • Mechanical operation
  • Ratchet thimble
  • IP54 water resistance rating
  • Hammertone finish
  • Includes plastic case
Catalog No. 50-005-33357 Supplier MSC Supplier No. 06233696
This mechanical blade micrometer offers precise measurements within a 1.0000 to 2.0000 inch range and an accuracy of 0.000150 " It features a ratchet thimble for consistent pressure and an IP54 rating for water resistance, making it suitable for various demanding environments.
  • Mechanical operation
  • Ratchet thimble
  • IP54 water resistance rating
  • Hammertone finish
  • Includes plastic case