Learn More
MSC Mitutoyo Borematic Replacement Head, 50-63mm Range, ±6μm Accuracy
Catalog No. 5000536282
Encompass
This Borematic replacement head is designed for precise internal diameter measurements within a 50-63mm range. It features durable titanium-coated measuring pins and an Absolute System for reliable and efficient operation, allowing for measurements even at the bottom of blind holes.
  • Titanium-coated measuring pins for excellent durability and impact resistance.
  • Allows measurement right to the bottom of a blind hole.
  • Digital display with quick-action lever for fast and easy measurements.
  • Built-in Absolute System eliminates origin point setting and prevents over-speed errors.
  • Compatible with statistical process control (SPC) systems.
  • Features a built-in tolerance judgment function for go/no-go inspections.
Catalog No. 50-005-36282 Supplier MSC Supplier No. 50830132
This Borematic replacement head is designed for precise internal diameter measurements within a 50-63mm range. It features durable titanium-coated measuring pins and an Absolute System for reliable and efficient operation, allowing for measurements even at the bottom of blind holes.
  • Titanium-coated measuring pins for excellent durability and impact resistance.
  • Allows measurement right to the bottom of a blind hole.
  • Digital display with quick-action lever for fast and easy measurements.
  • Built-in Absolute System eliminates origin point setting and prevents over-speed errors.
  • Compatible with statistical process control (SPC) systems.
  • Features a built-in tolerance judgment function for go/no-go inspections.