Learn More
MSC Mitutoyo Calibration Gage Set, For LSM-501S, 0.1 mm, 10 mm
Catalog No. 5000528071
Encompass
This calibration gage set is designed for use with the LSM-501S laser scan micrometer. It provides precise 0.1 mm and 10 mm gages, along with a stand and carrying case, to ensure accurate calibration of compatible micrometers.
  • Standard cylinder gage set suitable for calibration of laser scan micrometers.
  • Compatible with specific laser scan micrometer models.
  • Includes 0.1 mm and 10 mm gages.
  • Comes with a stand and carrying case.
Catalog No. 50-005-28071 Supplier MSC Supplier No. 62439336
This calibration gage set is designed for use with the LSM-501S laser scan micrometer. It provides precise 0.1 mm and 10 mm gages, along with a stand and carrying case, to ensure accurate calibration of compatible micrometers.
  • Standard cylinder gage set suitable for calibration of laser scan micrometers.
  • Compatible with specific laser scan micrometer models.
  • Includes 0.1 mm and 10 mm gages.
  • Comes with a stand and carrying case.