Learn More
MSC Mitutoyo Caliper Base, 100 mm, for Depth Bar Calipers, with Depth Rod
Catalog No. 5000533771
Encompass
This caliper base is designed to convert standard calipers into depth gauges. It features an overall length of 100 mm and includes a depth rod, making it suitable for precise depth measurements with calipers that have A Depth Bar.
  • 1 piece construction
  • Includes depth rod
  • For use with calipers with A Depth Bar
  • Compatible with 4 in, 6 in, 8 in, 12 in / 100 mm, 150 mm, 200 mm, 300 mm vernier, dial and digital calipers
  • Finely grounded base surface
  • Secure locking clamp
Catalog No. 50-005-33771 Supplier MSC Supplier No. 35347707
This caliper base is designed to convert standard calipers into depth gauges. It features an overall length of 100 mm and includes a depth rod, making it suitable for precise depth measurements with calipers that have A Depth Bar.
  • 1 piece construction
  • Includes depth rod
  • For use with calipers with A Depth Bar
  • Compatible with 4 in, 6 in, 8 in, 12 in / 100 mm, 150 mm, 200 mm, 300 mm vernier, dial and digital calipers
  • Finely grounded base surface
  • Secure locking clamp