Learn More
MSC Mitutoyo Caliper Micrometer Kit, 4 pc, 0-6 in, 0-1 in
Catalog No. 5000531674
Encompass
This 4-piece Machinist Caliper & Micrometer Kit includes a Digimatic Caliper with Absolute Encoder and a Digimatic Micrometer. This comprehensive kit provides precise measurements with a caliper range of 0 to 6 inches and a micrometer range of 0 to 1 " Both instruments offer the flexibility of switching between imperial and metric units, making them versatile for various applications. The set is housed in a durable Mahogany Case and also includes a Micrometer Spanner.
  • Digimatic caliper with absolute encoder
  • Micrometer spanner included
  • Digimatic micrometer for precise measurements
  • Housed in a durable mahogany case
  • Caliper provides a resolution of 0.000005 inches
  • Caliper maximum measurement of 6 inches
  • Caliper minimum measurement of 0 inches
  • Micrometer provides a graduation of 0.001000 inches
  • Ability to switch between imperial and metric measurements
Catalog No. 50-005-31674 Supplier MSC Supplier No. 90918160
This 4-piece Machinist Caliper & Micrometer Kit includes a Digimatic Caliper with Absolute Encoder and a Digimatic Micrometer. This comprehensive kit provides precise measurements with a caliper range of 0 to 6 inches and a micrometer range of 0 to 1 " Both instruments offer the flexibility of switching between imperial and metric units, making them versatile for various applications. The set is housed in a durable Mahogany Case and also includes a Micrometer Spanner.
  • Digimatic caliper with absolute encoder
  • Micrometer spanner included
  • Digimatic micrometer for precise measurements
  • Housed in a durable mahogany case
  • Caliper provides a resolution of 0.000005 inches
  • Caliper maximum measurement of 6 inches
  • Caliper minimum measurement of 0 inches
  • Micrometer provides a graduation of 0.001000 inches
  • Ability to switch between imperial and metric measurements