Learn More
MSC Mitutoyo Depth Gage Attachment, 100 mm Length, 5.5 mm Minimum Hole, for Metric Height Gages
Catalog No. 5000529661
Encompass
This depth gage attachment is designed for use with height gages to accurately measure groove and hole depths. It features a metric-type holding bar and is compatible with standard dial indicator contact points for versatile applications.
  • Measures groove and hole depth when attached to a height gage
  • Minimum measurable hole diameter of 5.5 mm
  • Maximum distance from holding bar to contact point is 80 mm
  • Equipped with a 9x9 mm cross-section metric holding bar
  • Holding bar length of 100 mm
  • Compatible with standard dial indicator points
Catalog No. 50-005-29661 Supplier MSC Supplier No. 00150367
This depth gage attachment is designed for use with height gages to accurately measure groove and hole depths. It features a metric-type holding bar and is compatible with standard dial indicator contact points for versatile applications.
  • Measures groove and hole depth when attached to a height gage
  • Minimum measurable hole diameter of 5.5 mm
  • Maximum distance from holding bar to contact point is 80 mm
  • Equipped with a 9x9 mm cross-section metric holding bar
  • Holding bar length of 100 mm
  • Compatible with standard dial indicator points