Learn More
MSC Mitutoyo Depth Micrometer, 0-4 in, 4 Rods, 4 in Base, 0.001 in Graduation
Catalog No. 5000533301
Encompass
This mechanical depth micrometer is designed for precise depth measurements, offering a 0-4 inch range. It comes with 4 interchangeable rods to provide a wide measuring range. Features include a ratchet stop thimble for constant measuring force and a measuring rod clamp, ensuring consistent and accurate readings for various industrial and scientific applications.
  • Ø4 mm interchangeable rods with lapped measuring end
  • Rod length can be adjusted in 1 in or 25 mm increments
  • Ratchet stop provides constant measuring force
  • Measuring rod clamp for secure readings
  • Supplied in a fitted plastic case for protection
  • Satin chrome finish for durability
  • Hardened steel measuring face material for longevity
  • High accuracy of 0.00012 in
Catalog No. 50-005-33301 Supplier MSC Supplier No. 06234454
This mechanical depth micrometer is designed for precise depth measurements, offering a 0-4 inch range. It comes with 4 interchangeable rods to provide a wide measuring range. Features include a ratchet stop thimble for constant measuring force and a measuring rod clamp, ensuring consistent and accurate readings for various industrial and scientific applications.
  • Ø4 mm interchangeable rods with lapped measuring end
  • Rod length can be adjusted in 1 in or 25 mm increments
  • Ratchet stop provides constant measuring force
  • Measuring rod clamp for secure readings
  • Supplied in a fitted plastic case for protection
  • Satin chrome finish for durability
  • Hardened steel measuring face material for longevity
  • High accuracy of 0.00012 in