Learn More
MSC Mitutoyo Depth Micrometer, 0-6 inch Range, 0.001 inch Graduation, 4 inch Base
Catalog No. 5000529297
Encompass
The Mitutoyo depth micrometer is designed for precise measurement of various depths. It features interchangeable rods with lapped measuring ends and a ratchet stop mechanism for consistent measuring force and improved repeatability. This tool is supplied with a fitted plastic case for protection and easy storage. This micrometer offers high accuracy for a wide range of applications.
  • Interchangeable rods with lapped measuring ends
  • Adjustable rod length in 1 inch increments
  • Ratchet stop ensures constant measuring force and repeatability
  • Measuring rod clamp provides added stability
  • Supplied with a fitted plastic case
Catalog No. 50-005-29297 Supplier MSC Supplier No. 18992933
The Mitutoyo depth micrometer is designed for precise measurement of various depths. It features interchangeable rods with lapped measuring ends and a ratchet stop mechanism for consistent measuring force and improved repeatability. This tool is supplied with a fitted plastic case for protection and easy storage. This micrometer offers high accuracy for a wide range of applications.
  • Interchangeable rods with lapped measuring ends
  • Adjustable rod length in 1 inch increments
  • Ratchet stop ensures constant measuring force and repeatability
  • Measuring rod clamp provides added stability
  • Supplied with a fitted plastic case