Learn More
MSC Mitutoyo Detector, 0.75mN, 2μm Stylus Tip, Skidless, ISO/JIS Compliant
Catalog No. 5000534110
Encompass
The Mitutoyo 178-396-2 is a low-force detector designed for ultra-sensitive surface roughness measurement. It features a 0.75mN measuring force, a 2μm stylus tip with a 60-degree geometry, and includes a skidless nosepiece. This detector is compliant with ISO and JIS standards, making it suitable for a wide range of precision applications in various industries.
  • Low-force sensor for delicate or precision-finished surfaces
  • Compliant with ISO and JIS standards
  • Includes a skidless nosepiece and standard stylus
  • Wide-range, high-resolution detector
  • Accurate drive unit for high-accuracy measurement
  • Supports both skidded and skidless measurement methods
Catalog No. 50-005-34110 Supplier MSC Supplier No. 18678664
The Mitutoyo 178-396-2 is a low-force detector designed for ultra-sensitive surface roughness measurement. It features a 0.75mN measuring force, a 2μm stylus tip with a 60-degree geometry, and includes a skidless nosepiece. This detector is compliant with ISO and JIS standards, making it suitable for a wide range of precision applications in various industries.
  • Low-force sensor for delicate or precision-finished surfaces
  • Compliant with ISO and JIS standards
  • Includes a skidless nosepiece and standard stylus
  • Wide-range, high-resolution detector
  • Accurate drive unit for high-accuracy measurement
  • Supports both skidded and skidless measurement methods