Learn More
MSC Mitutoyo Dial Bore Gage, 1.4 to 2.5 in, 0.00008 in Accuracy
Catalog No. 5000532468
Encompass
This dial bore gage allows for high-accuracy inside diameter measurements. It features interchangeable anvils and durable carbide-tipped contact points. Designed with a large grip that includes a sponge-like structure, it effectively reduces heat transfer from the operator's hand, ensuring stable and precise readings.
  • Accurate inside diameter measurement
  • Extended effective stroke
  • Durable carbide-tipped contact points
  • Large grip reduces heat transfer for stable readings
  • Includes interchangeable washers for fine setting
  • Supports optional extension rods for deep holes
  • Indicator included
  • Accessories included
  • Two-point contact system
  • Pendulum guide structure for easy measurement
  • Dial gauge protector for durability
Catalog No. 50-005-32468 Supplier MSC Supplier No. 43300243
This dial bore gage allows for high-accuracy inside diameter measurements. It features interchangeable anvils and durable carbide-tipped contact points. Designed with a large grip that includes a sponge-like structure, it effectively reduces heat transfer from the operator's hand, ensuring stable and precise readings.
  • Accurate inside diameter measurement
  • Extended effective stroke
  • Durable carbide-tipped contact points
  • Large grip reduces heat transfer for stable readings
  • Includes interchangeable washers for fine setting
  • Supports optional extension rods for deep holes
  • Indicator included
  • Accessories included
  • Two-point contact system
  • Pendulum guide structure for easy measurement
  • Dial gauge protector for durability