Learn More
MSC Mitutoyo Dial Caliper, 0-300 mm, 0.02 mm Graduation, Stainless Steel
Catalog No. 5000510980
Encompass
This dial caliper offers precise measurements with a 0-300 mm range and 0.02 mm graduation. Constructed from hardened stainless steel, it provides durability and corrosion resistance for demanding applications, ensuring smooth sliding and high-shock protection.
  • Hardened stainless steel for durability and corrosion resistance.
  • New dial movement provides ultra-smooth sliding and high-shock protection.
  • Measures OD, ID, depth, and steps.
  • Thumb roller for consistent feel and accurate measurements.
  • Titanium nitride coated main beam sliding surfaces reduce wear.
  • Extra rigid main beam.
  • Micro-fine graduations.
  • Adjustable and locking bezel.
  • Covered rack keeps gear teeth clean.
  • Supplied in a fitted plastic case.
Catalog No. 50-005-10980 Supplier MSC Supplier No. 84477728
This dial caliper offers precise measurements with a 0-300 mm range and 0.02 mm graduation. Constructed from hardened stainless steel, it provides durability and corrosion resistance for demanding applications, ensuring smooth sliding and high-shock protection.
  • Hardened stainless steel for durability and corrosion resistance.
  • New dial movement provides ultra-smooth sliding and high-shock protection.
  • Measures OD, ID, depth, and steps.
  • Thumb roller for consistent feel and accurate measurements.
  • Titanium nitride coated main beam sliding surfaces reduce wear.
  • Extra rigid main beam.
  • Micro-fine graduations.
  • Adjustable and locking bezel.
  • Covered rack keeps gear teeth clean.
  • Supplied in a fitted plastic case.