This Mitutoyo dial caliper offers a 6-inch measurement capacity with a 0.001 inch graduation and an accuracy of 0.001 " It measures 0.1 inch per revolution and is calibrated. The caliper features durable carbide-tipped jaws, making it suitable for depth, inside, outside, and step measurements.
- 0.001 inch accuracy.
- 6 inch maximum measurement.
- 0.1 inch per revolution range.
- Sliding jaw adjustment.
- Carbide-tipped jaws for durability.
- Supports inside and outside jaw measurements.
- 0.001 inch dial graduation.
- NIST traceability certified.
- Pre-calibrated for immediate use.
- Comes in a fitted plastic case.
- Improved finish on sliding surfaces for longevity.
- New font for easier readability.
- Lock screw for dial bezel and for holding the sliding jaw position.
- Ultra smooth sliding and high shock protection.