Learn More
MSC Mitutoyo Dial Caliper, 6 in Range, 0.001 in Graduation, Green Face
Supplier: MSC 84477769
This dial caliper features hardened stainless steel construction for durability and corrosion resistance. It offers versatile measurement capabilities, including outside, inside, step, and depth measurements, making it suitable for various applications. The design incorporates a thumb roller for consistent, accurate readings and a rigid main beam.
- Hardened stainless steel for longer life and corrosion resistance
- Thumb roller provides consistent feel for accurate measurements
- Performs outside dimension (OD), inside dimension (ID), step, and depth measurements
- Sliding surfaces of the main beam are TiN coated to decrease wear
- Extra rigid main beam
- Micro-fine graduations
- Adjustable and locking bezel
- Covered rack to keep gear teeth clean
- Easy zero adjustment
- Finished with satin chrome
- Supplied in a fitted plastic case