Learn More
MSC Mitutoyo Dial Drop Indicator, 0.025 in Range, 0.0001 in Graduation
Catalog No. 5000513617
Encompass
This dial drop indicator provides precise measurements for various applications. It offers clear, accurate readings with robust design features like shockproof construction and jeweled bearings, ensuring durability and reliable performance.
  • Accuracy of ±0.0001 in.
  • Flat back, plunger indicator style, and jeweled bearing.
  • Measuring force of 2.0 N or less; range per revolution of 0.01 in.
  • Shockproof design with glare-free, scratch-resistant crystal.
  • Meets ANSI/AGD and ISO/JIS standards.
  • Carbide tipped contact point for durability.
  • Enhanced resistance to dust, water, and oil.
  • Four-screw mounting; machine ground rack gear for accuracy.
  • Includes a rotating bezel.
  • Hardened steel spindle/stem with offset bushing for clamping.
Catalog No. 50-005-13617 Supplier MSC Supplier No. 29454709
This dial drop indicator provides precise measurements for various applications. It offers clear, accurate readings with robust design features like shockproof construction and jeweled bearings, ensuring durability and reliable performance.
  • Accuracy of ±0.0001 in.
  • Flat back, plunger indicator style, and jeweled bearing.
  • Measuring force of 2.0 N or less; range per revolution of 0.01 in.
  • Shockproof design with glare-free, scratch-resistant crystal.
  • Meets ANSI/AGD and ISO/JIS standards.
  • Carbide tipped contact point for durability.
  • Enhanced resistance to dust, water, and oil.
  • Four-screw mounting; machine ground rack gear for accuracy.
  • Includes a rotating bezel.
  • Hardened steel spindle/stem with offset bushing for clamping.