Learn More
MSC Mitutoyo Dial Drop Indicator, 0.5000" Max Measurement, 0.000500" Accuracy
Catalog No. 5000530324
Encompass
This dial drop indicator offers a 0.5000 inch maximum measurement and a 0-50 dial reading, providing high accuracy of 0.000500 inch for precise measurements.
  • White dial
  • 0.000500 inch dial graduation
  • 0.0500 inch per revolution range
  • Flat back design
  • Includes a revolution counter
  • 0.5000 inch maximum measurement
  • 0.000500 inch accuracy
  • 0-50 Dial reading
  • 1.80 N measuring force
  • 57.00 mm dial diameter
  • 3/8 inch stem diameter
  • 103.00 mm overall length
Catalog No. 50-005-30324 Supplier MSC Supplier No. 64489529
This dial drop indicator offers a 0.5000 inch maximum measurement and a 0-50 dial reading, providing high accuracy of 0.000500 inch for precise measurements.
  • White dial
  • 0.000500 inch dial graduation
  • 0.0500 inch per revolution range
  • Flat back design
  • Includes a revolution counter
  • 0.5000 inch maximum measurement
  • 0.000500 inch accuracy
  • 0-50 Dial reading
  • 1.80 N measuring force
  • 57.00 mm dial diameter
  • 3/8 inch stem diameter
  • 103.00 mm overall length