Learn More
MSC Mitutoyo Dial Height Gage, 0-24 in, 0.001 in Graduation, 0.002 in Accuracy
Catalog No. 5000533429
Encompass
The Mitutoyo 192-152 Dial Height Gage provides accurate, easy-to-read fine and coarse measurements using a dial and counter display. It ensures error-free reading with dual digital counters and features a 0-24 inch range, 0.001 inch graduation, and 0.002 inch accuracy. A carbide-tipped scriber and hand wheel are included for precise operation.
  • Dial and counter display provides accurate and easy-to-read measurements.
  • Dual digital counters allow resettable upward and downward measurements.
  • Includes a durable carbide-tipped scriber.
  • Hand wheel and feed wheel facilitate precise adjustments.
  • Counters and dial can be re-zeroed at any scriber position.
  • Rigid, double-column structure ensures high accuracy and repeatability.
  • Base is shaped for a secure grip.
  • Ergonomic inclined slider handle.
Catalog No. 50-005-33429 Supplier MSC Supplier No. 06243257
The Mitutoyo 192-152 Dial Height Gage provides accurate, easy-to-read fine and coarse measurements using a dial and counter display. It ensures error-free reading with dual digital counters and features a 0-24 inch range, 0.001 inch graduation, and 0.002 inch accuracy. A carbide-tipped scriber and hand wheel are included for precise operation.
  • Dial and counter display provides accurate and easy-to-read measurements.
  • Dual digital counters allow resettable upward and downward measurements.
  • Includes a durable carbide-tipped scriber.
  • Hand wheel and feed wheel facilitate precise adjustments.
  • Counters and dial can be re-zeroed at any scriber position.
  • Rigid, double-column structure ensures high accuracy and repeatability.
  • Base is shaped for a secure grip.
  • Ergonomic inclined slider handle.