Learn More
MSC Mitutoyo Dial Indicator, 0.125 in, 0.0005 in Graduation, 0-50 Dial Reading, Lug Back
Catalog No. 5000527713
Encompass
This Standard Type Dial Indicator provides excellent accuracy and reliability for various applications. It features a hardened, high-strength steel stem and spindle, with jeweled bearings for smooth, precise operations. Protected against water and oil, its crystal has a scratch and glare-resistant coating for durability and readability. It conforms to ANSI/AGD Standards.
  • Available in Inch models only.
  • Offered with lug back or flat back for mounting purposes.
  • Unique shock-proof mechanism protects against shock from sudden spindle retraction.
  • Rack gear is machine ground for tighter fit, increased accuracy.
  • Contact point is carbide tipped for lasting accuracy, durability.
  • Crystal is glare-free with a scratch-resistant coating.
  • Supplied in a fitted plastic case.
Catalog No. 50-005-27713 Supplier MSC Supplier No. 71458301
This Standard Type Dial Indicator provides excellent accuracy and reliability for various applications. It features a hardened, high-strength steel stem and spindle, with jeweled bearings for smooth, precise operations. Protected against water and oil, its crystal has a scratch and glare-resistant coating for durability and readability. It conforms to ANSI/AGD Standards.
  • Available in Inch models only.
  • Offered with lug back or flat back for mounting purposes.
  • Unique shock-proof mechanism protects against shock from sudden spindle retraction.
  • Rack gear is machine ground for tighter fit, increased accuracy.
  • Contact point is carbide tipped for lasting accuracy, durability.
  • Crystal is glare-free with a scratch-resistant coating.
  • Supplied in a fitted plastic case.