Learn More
MSC Mitutoyo Dial Snap Gage, 1 to 2 inch, Solid Carbide Anvil
Catalog No. 5000516773
Encompass
The Mitutoyo Dial Snap Gage is engineered for rapid GO/NO-GO assessment of cylindrical and shaft diameters. This durable gage offers a 1 to 2 inch measuring range and is equipped with solid carbide anvils, ensuring consistent and accurate results. Constructed with a robust steel frame, it is designed to accept a standard dial indicator (sold separately) with a 3/8 inch stem.
  • Quickly judges GO/NO-GO for cylinders and shafts
  • Wide, flat carbide anvils for enhanced stability
  • Adjustable measuring range for versatility
  • Features a 0.078 inch (2 mm) anvil retracting stroke
  • Ideal for use in machining processes
  • Robust steel frame construction
  • Compatible with indicators that have a 3/8 inch stem
Catalog No. 50-005-16773 Supplier MSC Supplier No. 06244099
The Mitutoyo Dial Snap Gage is engineered for rapid GO/NO-GO assessment of cylindrical and shaft diameters. This durable gage offers a 1 to 2 inch measuring range and is equipped with solid carbide anvils, ensuring consistent and accurate results. Constructed with a robust steel frame, it is designed to accept a standard dial indicator (sold separately) with a 3/8 inch stem.
  • Quickly judges GO/NO-GO for cylinders and shafts
  • Wide, flat carbide anvils for enhanced stability
  • Adjustable measuring range for versatility
  • Features a 0.078 inch (2 mm) anvil retracting stroke
  • Ideal for use in machining processes
  • Robust steel frame construction
  • Compatible with indicators that have a 3/8 inch stem