Learn More
MSC Mitutoyo Dial Test Indicator, Horizontal, 0.016 inch Range, 0.0001 inch Graduation
Catalog No. 5000514037
Encompass
This Dial Test Indicator Full Set provides precise measurements in difficult-to-access areas. Its unique horizontal 20° tilted face ensures easy reading, ideal for large workpieces or tall workbenches. With a reversible measuring direction and adjustable contact point, it offers high-precision measurement of surface variations. Its durable design includes high-impact resistance and jeweled bearings for quick, sensitive response.
  • Horizontal 20° tilted face for easy viewing.
  • No-clutch structure allows automatic reversal of measuring direction.
  • O-ring seal and bonded crystal protect against dust and oil.
  • Carbide-tipped contact point for lasting accuracy.
  • Glare-free, scratch-resistant crystal.
  • Full set includes multiple contact points and holding bar.
Catalog No. 50-005-14037 Supplier MSC Supplier No. 06385488
This Dial Test Indicator Full Set provides precise measurements in difficult-to-access areas. Its unique horizontal 20° tilted face ensures easy reading, ideal for large workpieces or tall workbenches. With a reversible measuring direction and adjustable contact point, it offers high-precision measurement of surface variations. Its durable design includes high-impact resistance and jeweled bearings for quick, sensitive response.
  • Horizontal 20° tilted face for easy viewing.
  • No-clutch structure allows automatic reversal of measuring direction.
  • O-ring seal and bonded crystal protect against dust and oil.
  • Carbide-tipped contact point for lasting accuracy.
  • Glare-free, scratch-resistant crystal.
  • Full set includes multiple contact points and holding bar.