Learn More
MSC Mitutoyo Dial Test Indicator, Horizontal, 0.50 mm Max, 0.01 mm Accuracy, Jeweled
Catalog No. 5000512161
Encompass
This dial test indicator is designed for precise horizontal measurements, offering a maximum measurement capacity of 0.50 mm and an accuracy of 0.01 mm. It features a jeweled bearing for smooth operation and a clear dial for easy reading.
  • Dust and oil proof construction
  • O-ring seal for enhanced protection
  • Carbide-tipped contact point for durability
  • Glare-free crystal ensures clear visibility
  • No-clutch structure for reliable performance
  • Automatic reversal for convenience
  • High-impact resistance for demanding environments
  • Durable and scratch-resistant design
Catalog No. 50-005-12161 Supplier MSC Supplier No. 62689815
This dial test indicator is designed for precise horizontal measurements, offering a maximum measurement capacity of 0.50 mm and an accuracy of 0.01 mm. It features a jeweled bearing for smooth operation and a clear dial for easy reading.
  • Dust and oil proof construction
  • O-ring seal for enhanced protection
  • Carbide-tipped contact point for durability
  • Glare-free crystal ensures clear visibility
  • No-clutch structure for reliable performance
  • Automatic reversal for convenience
  • High-impact resistance for demanding environments
  • Durable and scratch-resistant design