Learn More
MSC Mitutoyo Dial Test Indicator Kit, 0.06 in Range, 0.0005 in Graduation
Catalog No. 5000529856
Encompass
This 9-piece Mitutoyo Dial Test Indicator Kit is a high-precision measurement tool for industrial and laboratory use. It features a 0.06 in measurement range, 0.0005 in graduation, and a 0-15-0 dial reading. This horizontal indicator kit offers durability, accuracy, and versatile components.
  • 0.06 in maximum measurement range
  • 0.0005 in graduation
  • 0-15-0 dial reading
  • Horizontal indicator style
  • O-ring seal and glare-free crystal
  • No-clutch structure with adjustable contact point
  • High-impact resistance and durable construction
  • Tilted face for easy reading
  • Reversible measuring direction and high sensitivity
  • Universal holder
  • Includes indicator, swivel clamp, holding bar, knurled clamp ring, contact points, and stems.
Catalog No. 50-005-29856 Supplier MSC Supplier No. 06385462
This 9-piece Mitutoyo Dial Test Indicator Kit is a high-precision measurement tool for industrial and laboratory use. It features a 0.06 in measurement range, 0.0005 in graduation, and a 0-15-0 dial reading. This horizontal indicator kit offers durability, accuracy, and versatile components.
  • 0.06 in maximum measurement range
  • 0.0005 in graduation
  • 0-15-0 dial reading
  • Horizontal indicator style
  • O-ring seal and glare-free crystal
  • No-clutch structure with adjustable contact point
  • High-impact resistance and durable construction
  • Tilted face for easy reading
  • Reversible measuring direction and high sensitivity
  • Universal holder
  • Includes indicator, swivel clamp, holding bar, knurled clamp ring, contact points, and stems.