Learn More
MSC Mitutoyo Digimatic Indicator, 0-2 in Range, 0.00002 in, Flat Back
Catalog No. 5000537580
Encompass
This Digimatic indicator offers a 0-2 in (0-50.8 mm) measuring range with selectable resolutions down to 0.00002 in. It features a large, easy-to-read LCD and an ABSOLUTE encoder. Designed with a flat back, it is suitable for various industrial measurements and includes SPC output.
  • Range: 0-2 in (0-50.8 mm)
  • Selectable resolution from 0.00002 in
  • Accuracy: +/- 0.00012 in
  • Repeatability: 0.00008
  • Large, easy-to-read LCD display
  • ABSOLUTE encoder for maintained settings
  • Preset, Zero-Set, inch/mm, +/- direction, and GO/±NG functions
  • Calibration due date alarm
  • Flat back design
  • SPC output
Catalog No. 50-005-37580 Supplier MSC Supplier No. 30007892
This Digimatic indicator offers a 0-2 in (0-50.8 mm) measuring range with selectable resolutions down to 0.00002 in. It features a large, easy-to-read LCD and an ABSOLUTE encoder. Designed with a flat back, it is suitable for various industrial measurements and includes SPC output.
  • Range: 0-2 in (0-50.8 mm)
  • Selectable resolution from 0.00002 in
  • Accuracy: +/- 0.00012 in
  • Repeatability: 0.00008
  • Large, easy-to-read LCD display
  • ABSOLUTE encoder for maintained settings
  • Preset, Zero-Set, inch/mm, +/- direction, and GO/±NG functions
  • Calibration due date alarm
  • Flat back design
  • SPC output