Learn More
MSC Mitutoyo MDH-25MB High-Accuracy Digimatic Micrometer, 0.5 μm Accuracy, 0.1 μm Resolution, 1 ea
Catalog No. 5000510344
Encompass
This high-accuracy Digimatic micrometer provides ±0.5 μm accuracy at 0.1 μm resolution, designed for rapid and reliable measurement of workpieces requiring superior precision. It features an integrated constant-force mechanism in the thimble and an absolute linear scale that eliminates the need for repeated origin point setting.
  • Equipped with an extremely rigid frame and a removable heat shield
  • Adjustable measuring force ranges from 0.5 N to 2.5 N
  • Fast spindle feed rate of 10 mm per rotation
  • Supports real-time data collection and statistical analysis via MeasurLink®
  • Extended battery life of approximately 5 years under normal usage
  • Carbide-tipped measuring faces for increased durability
  • Integrated function lock prevents accidental origin changes during use
Catalog No. 50-005-10344 Supplier MSC Supplier No. 06389514
This high-accuracy Digimatic micrometer provides ±0.5 μm accuracy at 0.1 μm resolution, designed for rapid and reliable measurement of workpieces requiring superior precision. It features an integrated constant-force mechanism in the thimble and an absolute linear scale that eliminates the need for repeated origin point setting.
  • Equipped with an extremely rigid frame and a removable heat shield
  • Adjustable measuring force ranges from 0.5 N to 2.5 N
  • Fast spindle feed rate of 10 mm per rotation
  • Supports real-time data collection and statistical analysis via MeasurLink®
  • Extended battery life of approximately 5 years under normal usage
  • Carbide-tipped measuring faces for increased durability
  • Integrated function lock prevents accidental origin changes during use