Learn More
Grainger MITUTOYO Direct Input Tool Digimatic
Catalog No. NC2395972
Encompass_Preferred
The Mitutoyo Direct Input Tool Digimatic Micrometer is a precision measuring instrument for measuring small dimensions. It has a measuring range of 0-25mm/0-1in with a resolution of 0.001mm/0.00005in. The micrometer has a ratchet stop for consistent measurements and advanced ABSOLUTE linear encoder technology for resistance to contamination and shocks. It can take measurements in metric or inch units. The micrometer comes with a certificate of inspection traceable to NIST. It is made in Japan by Mitutoyo, a leading manufacturer of precision measuring tools.
Catalog No. NC2395972 Supplier Grainger Supplier No. 3VYZ1
Add to Cart
Add to Cart
The Mitutoyo Direct Input Tool Digimatic Micrometer is a precision measuring instrument for measuring small dimensions. It has a measuring range of 0-25mm/0-1in with a resolution of 0.001mm/0.00005in. The micrometer has a ratchet stop for consistent measurements and advanced ABSOLUTE linear encoder technology for resistance to contamination and shocks. It can take measurements in metric or inch units. The micrometer comes with a certificate of inspection traceable to NIST. It is made in Japan by Mitutoyo, a leading manufacturer of precision measuring tools.