Learn More
MSC Mitutoyo Drop Indicator Backs, Flat Back, AGD Group 1
Catalog No. 5000513380
Encompass
Mitutoyo drop indicator backs are designed to replace the original back of an indicator, particularly for special applications. This flat back type is compatible with AGD Group 1 indicators, ensuring precise fit and functionality.
  • Flat back type
  • Compatible with AGD Group 1 indicators
  • Suitable for special applications where the indicator back needs replacement
  • Made in Japan
Catalog No. 50-005-13380 Supplier MSC Supplier No. 01692722
Mitutoyo drop indicator backs are designed to replace the original back of an indicator, particularly for special applications. This flat back type is compatible with AGD Group 1 indicators, ensuring precise fit and functionality.
  • Flat back type
  • Compatible with AGD Group 1 indicators
  • Suitable for special applications where the indicator back needs replacement
  • Made in Japan