Learn More
MSC Mitutoyo Drop Indicator Flat Contact Points, #4-48 Thread, 0.3750 in, Solid Carbide
Catalog No. 5000527532
Encompass
Mitutoyo Drop Indicator Flat Contact Points are designed for precise measurement applications. These durable contact points feature a flat style, a #4-48 thread size, and are constructed from solid carbide for extended tool life and accuracy. They are compatible with AGD 1, 2, 3, and 4 dial gages.
  • Flat contact point style
  • Solid carbide point material
  • Contact point length: 0.3750 in
  • #4-48 thread size
  • Compatible with AGD 1, 2, 3, and 4 dial gages
  • Suitable for mounting on dial gages
Catalog No. 50-005-27532 Supplier MSC Supplier No. 77027100
Mitutoyo Drop Indicator Flat Contact Points are designed for precise measurement applications. These durable contact points feature a flat style, a #4-48 thread size, and are constructed from solid carbide for extended tool life and accuracy. They are compatible with AGD 1, 2, 3, and 4 dial gages.
  • Flat contact point style
  • Solid carbide point material
  • Contact point length: 0.3750 in
  • #4-48 thread size
  • Compatible with AGD 1, 2, 3, and 4 dial gages
  • Suitable for mounting on dial gages