Learn More
MSC Mitutoyo Drop Indicator Flat Contact Points, #4-48 Thread, 0.3750 in Point Length, Solid Carbide
Catalog No. 5000533994
Encompass
This Mitutoyo drop indicator flat contact point is designed for precision measurements. Made from solid carbide, it features a #4-48 thread and a 0.3750 in point length, making it compatible with various AGD series indicators.
  • Flat contact point style.
  • Solid carbide material for durability.
  • 0.3750 in point length.
  • #4-48 thread size.
  • Compatible with AGD 1, 2, 3, and 4 series indicators.
Catalog No. 50-005-33994 Supplier MSC Supplier No. 32814253
This Mitutoyo drop indicator flat contact point is designed for precision measurements. Made from solid carbide, it features a #4-48 thread and a 0.3750 in point length, making it compatible with various AGD series indicators.
  • Flat contact point style.
  • Solid carbide material for durability.
  • 0.3750 in point length.
  • #4-48 thread size.
  • Compatible with AGD 1, 2, 3, and 4 series indicators.