Learn More
MSC Mitutoyo Electronic Caliper, 0 to 18 in, 0.0005 in Resolution, Stainless Steel
Catalog No. 5000516927
Encompass
This electronic caliper provides a maximum measurement of 450.00 mm or 18 inches. It offers a resolution of 0.01 mm (0.0005 in) and an accuracy of +/-0.05 mm (+/-0.0020 in). The tool is made of stainless steel and is equipped with data output functionalities, making it suitable for measuring inside diameter, outside diameter, and step.
  • Data output capability.
  • Constructed from stainless steel.
  • Capable of measuring inside diameter, outside diameter, and step.
  • Features a hardened stainless steel finish.
  • Includes inside diameter jaws, a battery, and a case.
  • TUV Rheinland certified.
  • Utilizes a sliding jaw adjustment type.
  • Powered by a silver oxide battery (SR44).
Catalog No. 50-005-16927 Supplier MSC Supplier No. 06257885
This electronic caliper provides a maximum measurement of 450.00 mm or 18 inches. It offers a resolution of 0.01 mm (0.0005 in) and an accuracy of +/-0.05 mm (+/-0.0020 in). The tool is made of stainless steel and is equipped with data output functionalities, making it suitable for measuring inside diameter, outside diameter, and step.
  • Data output capability.
  • Constructed from stainless steel.
  • Capable of measuring inside diameter, outside diameter, and step.
  • Features a hardened stainless steel finish.
  • Includes inside diameter jaws, a battery, and a case.
  • TUV Rheinland certified.
  • Utilizes a sliding jaw adjustment type.
  • Powered by a silver oxide battery (SR44).