Learn More
MSC Mitutoyo Electronic Caliper, 0-8 in., 0.0005 in. Resolution, IP67
Catalog No. 5000534176
Encompass
This electronic caliper offers high-precision measurements in harsh environments. With IP67 protection against coolants, dust, and temporary water submersion, it ensures reliable operation. Its ABSOLUTE measurement system eliminates origin reset, allowing immediate use.
  • IP67 protection against coolants, dust, and temporary water submersion.
  • Includes certification for IP compliance.
  • Oil resistant materials used in all plastic components.
  • Incorporates Absolute measurement system; no origin reset needed.
  • SPC data output enables connection to measurement systems.
  • Approximately 5-year extended battery life.
  • Automatic power-on/off function.
  • Display readings increased by 22% for improved visibility.
  • Advanced ergonomic design for ease of use.
  • Capable of measuring stepped features.
Catalog No. 50-005-34176 Supplier MSC Supplier No. 00315911
This electronic caliper offers high-precision measurements in harsh environments. With IP67 protection against coolants, dust, and temporary water submersion, it ensures reliable operation. Its ABSOLUTE measurement system eliminates origin reset, allowing immediate use.
  • IP67 protection against coolants, dust, and temporary water submersion.
  • Includes certification for IP compliance.
  • Oil resistant materials used in all plastic components.
  • Incorporates Absolute measurement system; no origin reset needed.
  • SPC data output enables connection to measurement systems.
  • Approximately 5-year extended battery life.
  • Automatic power-on/off function.
  • Display readings increased by 22% for improved visibility.
  • Advanced ergonomic design for ease of use.
  • Capable of measuring stepped features.