Learn More
MSC Mitutoyo Electronic Caliper, 150 mm, 0.01 mm Resolution, Coolant-Proof
Catalog No. 5000514505
Encompass
This electronic caliper is constructed from hardened stainless steel and features carbide-tipped jaws, offering precise measurements up to 150 mm (6 inches) with a resolution of 0.01 mm (0.0005 inch) and an accuracy of 0.02 mm (0.0010 inch). It is coolant-proof with an IP67 rating, making it suitable for demanding workshop environments. The caliper supports data output and is ideal for inside, outside, step, and depth measurements.
  • Hardened stainless steel construction
  • Carbide-tipped jaws
  • Measurement range up to 150 mm (6 in) with 0.01 mm (0.0005 in) resolution and 0.02 mm (0.0010 in) accuracy
  • IP67 coolant-proof rating
  • Data output capabilities
  • Suitable for inside diameter, step, outside diameter, and depth measurements
  • Includes inside diameter jaws, depth measurement rod, and case
  • Powered by a single SR44 silver oxide battery
  • TUV Rheinland certified
Catalog No. 50-005-14505 Supplier MSC Supplier No. 84177666
This electronic caliper is constructed from hardened stainless steel and features carbide-tipped jaws, offering precise measurements up to 150 mm (6 inches) with a resolution of 0.01 mm (0.0005 inch) and an accuracy of 0.02 mm (0.0010 inch). It is coolant-proof with an IP67 rating, making it suitable for demanding workshop environments. The caliper supports data output and is ideal for inside, outside, step, and depth measurements.
  • Hardened stainless steel construction
  • Carbide-tipped jaws
  • Measurement range up to 150 mm (6 in) with 0.01 mm (0.0005 in) resolution and 0.02 mm (0.0010 in) accuracy
  • IP67 coolant-proof rating
  • Data output capabilities
  • Suitable for inside diameter, step, outside diameter, and depth measurements
  • Includes inside diameter jaws, depth measurement rod, and case
  • Powered by a single SR44 silver oxide battery
  • TUV Rheinland certified