Learn More
MSC Mitutoyo Electronic Caliper, 150 mm, 0.01 mm Resolution, IP67, Carbide-Tipped
Catalog No. 5000528199
Encompass
This electronic caliper is engineered for precise measurements in demanding industrial environments. With an IP67 rating for protection against coolants, water, dust, and oil, and durable carbide-tipped jaws, it offers versatile functionality for inside, outside, step, and depth measurements.
  • ABSOLUTE measurement system, no origin reset.
  • IP67 dust and water protection.
  • Carbide-tipped jaws for wear resistance.
  • Measures inside, outside, step, and depth.
  • 5-year battery life.
  • Auto power-on/off, retains origin.
  • Large display for visibility.
  • Hardened stainless steel.
  • Includes battery and case.
  • Unlimited response speed.
  • TUV Rheinland certified.
Catalog No. 50-005-28199 Supplier MSC Supplier No. 35099696
Add to Cart
Add to Cart
This electronic caliper is engineered for precise measurements in demanding industrial environments. With an IP67 rating for protection against coolants, water, dust, and oil, and durable carbide-tipped jaws, it offers versatile functionality for inside, outside, step, and depth measurements.
  • ABSOLUTE measurement system, no origin reset.
  • IP67 dust and water protection.
  • Carbide-tipped jaws for wear resistance.
  • Measures inside, outside, step, and depth.
  • 5-year battery life.
  • Auto power-on/off, retains origin.
  • Large display for visibility.
  • Hardened stainless steel.
  • Includes battery and case.
  • Unlimited response speed.
  • TUV Rheinland certified.