Learn More
MSC Mitutoyo Electronic Caliper, 1500 mm, 0.01 mm, Carbon Fiber Reinforced Plastic
Catalog No. 5000515158
Encompass
This electronic caliper offers precise measurements up to 1500 mm with a 0.01 mm resolution. Constructed from carbon fiber reinforced plastic (CFRP), it is lightweight and durable. Designed for both inside and outside diameter measurements, this caliper also features SPC data output and an IP66 rating.
  • Lightweight design
  • Carbon fiber reinforced plastic (CFRP) in beam and jaws
  • Stainless steel expansion strip for enhanced durability
  • Large LCD display for easy reading
  • Direct readout of inside diameter measurements
  • Offset and preset functions for versatile use
  • SPC data output capability
Catalog No. 50-005-15158 Supplier MSC Supplier No. 90500331
This electronic caliper offers precise measurements up to 1500 mm with a 0.01 mm resolution. Constructed from carbon fiber reinforced plastic (CFRP), it is lightweight and durable. Designed for both inside and outside diameter measurements, this caliper also features SPC data output and an IP66 rating.
  • Lightweight design
  • Carbon fiber reinforced plastic (CFRP) in beam and jaws
  • Stainless steel expansion strip for enhanced durability
  • Large LCD display for easy reading
  • Direct readout of inside diameter measurements
  • Offset and preset functions for versatile use
  • SPC data output capability