Learn More
MSC Mitutoyo Electronic Caliper, 1500 mm Range, 0.01 mm Resolution, Carbon Fiber Reinforced Plastic
Catalog No. 5000514571
Encompass
This electronic caliper provides precise measurements for various applications. It offers a measurement range up to 1500 mm (60 in) with a high resolution of 0.01 mm (0.0005 in) and an accuracy of ±0.10 mm (±0.0040 in). Constructed from durable carbon fiber reinforced plastic, this caliper is designed for reliability in industrial settings.
  • Measures up to 1500 mm (60 in)
  • Resolution of 0.01 mm (0.0005 in)
  • Accuracy of ±0.10 mm (±0.0040 in)
  • Constructed from carbon fiber reinforced plastic
  • Features data output, compatible with SPC cable or wireless transmitter
  • IP66 rated for dust and strong water jet protection
  • Suitable for inside and outside diameter measurements
  • NIST traceability and TUV Rheinland certified
  • Includes battery and case
Catalog No. 50-005-14571 Supplier MSC Supplier No. 53766267
This electronic caliper provides precise measurements for various applications. It offers a measurement range up to 1500 mm (60 in) with a high resolution of 0.01 mm (0.0005 in) and an accuracy of ±0.10 mm (±0.0040 in). Constructed from durable carbon fiber reinforced plastic, this caliper is designed for reliability in industrial settings.
  • Measures up to 1500 mm (60 in)
  • Resolution of 0.01 mm (0.0005 in)
  • Accuracy of ±0.10 mm (±0.0040 in)
  • Constructed from carbon fiber reinforced plastic
  • Features data output, compatible with SPC cable or wireless transmitter
  • IP66 rated for dust and strong water jet protection
  • Suitable for inside and outside diameter measurements
  • NIST traceability and TUV Rheinland certified
  • Includes battery and case