Learn More
MSC Mitutoyo Electronic Caliper, IP67, 200 mm, Stainless Steel
Catalog No. 5000512897
Encompass
This electronic caliper offers IP67 protection against coolants, dust, and temporary water submersion. It features an Absolute measurement system for 'set and forget' operation and is constructed from hardened stainless steel. With a 200 mm (8 inch) measurement range, it handles inside, outside, step, and depth measurements.
  • IP67 protection against coolants, dust, and water submersion
  • Absolute measurement system with set and forget technology
  • Increased battery life and functionality from custom chip design
  • NIST traceable certification for accuracy
  • Automatic power-on/off
  • Capable of depth, ID, OD, and step measurements
  • Constructed from hardened stainless steel
  • Includes battery, case, ID jaws, and depth measurement rod
  • TUV Rheinland certified
Catalog No. 50-005-12897 Supplier MSC Supplier No. 09683533
This electronic caliper offers IP67 protection against coolants, dust, and temporary water submersion. It features an Absolute measurement system for 'set and forget' operation and is constructed from hardened stainless steel. With a 200 mm (8 inch) measurement range, it handles inside, outside, step, and depth measurements.
  • IP67 protection against coolants, dust, and water submersion
  • Absolute measurement system with set and forget technology
  • Increased battery life and functionality from custom chip design
  • NIST traceable certification for accuracy
  • Automatic power-on/off
  • Capable of depth, ID, OD, and step measurements
  • Constructed from hardened stainless steel
  • Includes battery, case, ID jaws, and depth measurement rod
  • TUV Rheinland certified