This electronic caliper is constructed from hardened stainless steel, offering IP67 protection against dust and temporary water immersion. It provides precise measurements for depth, inside and outside diameters, and steps, featuring an easy-to-read display and a 5-year battery life.
- IP67 rating for harsh workshop conditions.
- Measures depth, inside/outside diameter, and step.
- 22% larger display for improved visibility.
- Ergonomic design with simple one-button operation.
- ABSOLUTE measurement system eliminates origin resetting.
- Automatic power on/off with 5-year battery life.
- Integrates with statistical process control systems.
- Factory inspection certificate included.
- Calibrated; includes battery and case.
- Hardened stainless steel construction.
- Knife-edge jaw style.
- NIST traceability certification.