Learn More
MSC Mitutoyo Electronic Depth Gage, 0.0076 mm Accuracy, 200 mm, 0.001 mm Resolution
Catalog No. 5000530727
Encompass
The Mitutoyo ABSOLUTE Digimatic Depth Gage is designed for precise depth measurements, featuring a hardened, ground, and lapped base for exceptional flatness. This electronic depth gage offers reliable and consistent performance by maintaining its origin point once set for the entire battery life.
  • Bottom surface of the base is hardened, ground, and lapped for highest degree of flatness.
  • Can keep track of the origin point once set for the entire life of the battery.
  • Includes extension rod and battery.
Catalog No. 50-005-30727 Supplier MSC Supplier No. 90977828
The Mitutoyo ABSOLUTE Digimatic Depth Gage is designed for precise depth measurements, featuring a hardened, ground, and lapped base for exceptional flatness. This electronic depth gage offers reliable and consistent performance by maintaining its origin point once set for the entire battery life.
  • Bottom surface of the base is hardened, ground, and lapped for highest degree of flatness.
  • Can keep track of the origin point once set for the entire life of the battery.
  • Includes extension rod and battery.