Learn More
MSC Mitutoyo Electronic Depth Micrometer, 0-6 inch, 4 inch Base, 6 Rods
Catalog No. 5000529241
Encompass
This electronic depth micrometer provides precise measurements across a 0-6 inch (0-152.4 mm) range with a resolution of 0.00005 inch (0.001 mm). It features a 4 inch base and includes 6 interchangeable rods, suitable for various depth measurement tasks.
  • 4 inch base
  • 6 interchangeable rods
  • Rod diameter of 4 mm
  • Rod length adjustable in 1 inch (25 mm) increments
  • Measuring rod clamps ensure measurement stability
  • Ratchet stop thimble provides constant measuring force
  • SPC output function
  • Functions: origin set, zero setting, data hold, data output, function lock, 2 presets, and inch/mm conversion
  • Alarms for low voltage and counting value composition error
  • Steel measuring face material
  • Uses an SR44 battery
Catalog No. 50-005-29241 Supplier MSC Supplier No. 13433289
This electronic depth micrometer provides precise measurements across a 0-6 inch (0-152.4 mm) range with a resolution of 0.00005 inch (0.001 mm). It features a 4 inch base and includes 6 interchangeable rods, suitable for various depth measurement tasks.
  • 4 inch base
  • 6 interchangeable rods
  • Rod diameter of 4 mm
  • Rod length adjustable in 1 inch (25 mm) increments
  • Measuring rod clamps ensure measurement stability
  • Ratchet stop thimble provides constant measuring force
  • SPC output function
  • Functions: origin set, zero setting, data hold, data output, function lock, 2 presets, and inch/mm conversion
  • Alarms for low voltage and counting value composition error
  • Steel measuring face material
  • Uses an SR44 battery